Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iodomethylcyclopentane, 98%, stabilized

Catalog No. AC378640050
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle

CAS: 27935-87-1 | C6H11I | 210.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27935-87-1
Molecular Formula C6H11I
Molecular Weight (g/mol) 210.06
MDL Number MFCD03453190
InChI Key DUMSKQUKLVSSII-UHFFFAOYSA-N
Synonym iodomethyl cyclopentane, cyclopentylmethyl iodide, cyclopentane, iodomethyl, cyclopentyliodomethane, acmc-20akpz, cyclopentylmethyliodide, iodomethyl-cyclopentane, cyclopentanemethyl iodide
PubChem CID 2762531
IUPAC Name iodomethylcyclopentane
SMILES ICC1CCCC1

Specifications

Boiling Point 78.0°C (17.0 mmHg)
Color Brown or Colorless to Orange
Physical Form Solution
Quantity 5 g
Assay Percent Range 97.5% min. (GC)
Linear Formula (C5H9)CH2I
Flash Point 92°C
Solubility Information Solubility in water: immiscible
Formula Weight 210.05
Percent Purity 98%
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.5400 to 1.5440
Specific Gravity 1.6
Density 1.6000g/mL
Chemical Name or Material Iodomethylcyclopentane, Stabilized
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Recommended Storage May darken on storage

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.