Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Iodopentafluorobenzene

Catalog No. 1031365G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-313-65G 5 g
10-313-625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 10-313-65G Supplier Sigma Aldrich Supplier No. 1031365G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Iodopentafluorobenzene forms supramolecular complexes with aromatic electron donors by forming halogen bonds to form discrete heterodimeric aggregates
  • Iodopentafluorobenzene was used to study the formation of radical ions of iodopentafluorobenzene in aqueous solution
  • It was used as solvent in a study to determine singlet oxgen lifetimes from phosphorescence decays in halogen substituted perfluorinated solvents by infrared emission spectrometery
  • It has potential applications in plasma processing industry and in preparation of catalysts

Specifications

CAS 827-15-6
Density 2.204 g/mL (at 25°C (literature))
Boiling Point 161°C (lit.)
Molecular Formula C6F5I
Refractive Index n20/D 1.496 (literature)
Linear Formula C6F5I
MDL Number MFCD00001032
Quantity 5 g
Molecular Weight (g/mol) 293.96
Percent Purity 99%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.