Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iodoquinol, Powder, USP, 96-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18603496
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
25 kg
Packaging:
Amber Glass Bottle

IO115, 83-73-8, C9H5I2NO

Spectrum™ Chemical Iodoquinol, Powder, USP Also known as Diquinol and iodoxin, is used in the treatment of amoebiasis and as a luminal amebicide. The USP grade indicates it is graded suitable for personal care, cosmetic and pharmaceutical applications. All Spectrum Chemical manufactured USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 83-73-8
Molecular Formula C9H5I2NO
Molecular Weight (g/mol) 396.95
InChI Key UXZFQZANDVDGMM-UHFFFAOYSA-N
IUPAC Name 5,7-diiodoquinolin-8-ol
SMILES OC1=C2N=CC=CC2=C(I)C=C1I

Specifications

CAS Max % 100%
Ignition Residue 0.5%
Loss on Drying 0.5%
Packaging Amber Glass Bottle
Quantity 100 g
Percent Purity 96 to 100.5%
Grade USP
Physical Form Crystalline Powder or Lumps
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.