Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ionic Strength Adjusting Buffer Ammonium Hydroxide 10% v/v in Isopropanol, Reagents

Catalog No. 01334953
Click to view available options
Quantity:
10 L

Ionic Strength Adjusting Buffer Ammonium Hydroxide 10% v/v in Isopropanol, Reagent, use for Water analysis. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Ionic Strength Adjusting Buffer Ammonium Hydroxide 10% v/v in Isopropanol
CAS 67-63-0, 1336-21-6
Boiling Point 100°C
Color Colorless
For Use With (Application) Water Analysis
Physical Form Liquid
Quantity 10 L
Solution Type Buffer
Packaging Cubitainer
Specific Gravity 0.7927
Sterility Non-sterile
Grade Reagent
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.