Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ionic Strength Adjustor for Sodium Testing, Reagents

Catalog No. 01334925
Click to view available options
Quantity:
500 mL
1 L
4 L

Catalog No. 01-334-925

Supplier: Reagents Holdings Llc CI180000500A

Only null left
Add to Cart
Add to Cart

Ionic Strength Adjustor for Sodium Testing, Reagent, use for Sodium analysis. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5, 12125-02-9
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
IUPAC Name water
SMILES O

Specifications

Packaging Poly Bottle
Specific Gravity 1.03
Physical Form Liquid
Color Colorless
Boiling Point 100°C
For Use With (Equipment) Water Analyzer
Solubility Completely miscible
Industry Type Sodium Analysis
Quantity 500 mL
Grade Reagent
Chemical Name or Material Ionic Strength Adjustor for Sodium Testing
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.