Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ionic Strength Adjustor, pH Indicating and Adjusting, for use with Ammonia Selective Electrode, Ricca Chemical
SDP

Catalog No. p-214924 Shop All Ricca Chemical Company Products
Click to view available options
Quantity:
500 mL
1 L

Catalog No. 4128-16

Supplier: Ricca Chemical Company 412816

Only null left
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Ionic Strength Adjustor
Name Note pH Indicating and Adjusting, for use with Ammonia Selective Electrode
CAS 7732-18-5, 1310-73-2, 67-56-1, 6381-92-6, 125-20-2
CAS Min % 65.0, 26.25, 5.14, 1.22, 0.01
CAS Max % 68.0, 27.32, 5.35, 1.27, 0.01
Grade Laboratory
Packaging Natural Poly Bottle
pH 13 to 14
Quantity 500 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.