Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ionomycin (Calcium Salt), Fisher BioReagents™

Catalog No. p-26587 Shop All Fisherbrand Products
Click to view available options
Quantity:
1 mg
Packaging:
Poly Micro Tube

Ionomycin (Calcium Salt)

  •  Suitable for use in electrophoretic separations, specifically polyacrylamide gel electrophoresis (PAGE)
  • This product is a 40% (w/v) solution of high quality acrylamide
  • Free of DNase, RNase, and protease
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56092-82-1
Molecular Formula C41H70CaO9
Molecular Weight (g/mol) 747.08
MDL Number MFCD00083634
InChI Key RULWQOBWZOQCOY-SQNLDGQHSA-M
PubChem CID 71311740
IUPAC Name calcium;(4R,6S,8S,10E,12R,14R,16E,18R,19R,20S,21S)-19,21-dihydroxy-22-[(2S,5S)-5-[(2R,5S)-5-[(1R)-1-hydroxyethyl]-5-methyloxolan-2-yl]-5-methyloxolan-2-yl]-4,6,8,12,14,18,20-heptamethyl-11-oxido-9-oxodocosa-10,16-dienoate
SMILES [Ca++].C[C@H](CCC([O-])=O)C[C@H](C)C[C@H](C)C(=O)[CH-]C(=O)[C@H](C)C[C@H](C)C\C=C\[C@@H](C)[C@@H](O)[C@@H](C)[C@@H](O)C[C@@H]1CC[C@](C)(O1)[C@H]1CC[C@](C)(O1)[C@@H](C)O

Specifications

Type Ionomycin Calcium Salt
Physical Form Solid
Melting Point 205°C to 206°C
Packaging Poly Micro Tube
Color White
Quantity 1 mg
Assay Percent Range ≥98.0 %
Formula Weight 747.1
Percent Purity 98.0% min. (by NP TLC), 99.0% min. (by RP HPLC)
Chemical Name or Material Ionomycin, Calcium Salt, From Streptomyces conglobatus
Recommended Storage 2 to 8°C, protect from light
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.