Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ IOX 2
GREENER_CHOICE

Catalog No. 445110
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-511-0 10 mg
44-515-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 44-511-0 Supplier Tocris Bioscience™ Supplier No. 4451/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent, selective HIF-1α prolyl hydroxylase-2 (PHD2) inhibitor

Potent inhibitor of HIF-1α prolyl hydroxylase-2 (PHD2) (IC50 = 21 nM). Displays over 100-fold selectivity for PHD2 over factor inhibiting HIF-1 (FIH-1) and histone demethylases JMJD2A, JMJD2C, JMJD2E and JMJD3. Cell permeable.

Chemical Identifiers

CAS 931398-72-0
Molecular Formula C19H16N2O5

Specifications

Quantity 10 mg
Formula Weight 352.34
Purity >99%
Molecular Weight (g/mol) 352.346
Chemical Name or Material IOX 2
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.