Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IP3 Receptor 1 Recombinant Rabbit Monoclonal Antibody (4Z1B6)
GREENER_CHOICE

Catalog No. PIMA535516
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA535516 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA535516 Supplier Invitrogen™ Supplier No. MA535516
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: HTAQIEIVRL DRTMEQIVFP VPSICEFLTK ESKLRIYYTT ERDEQGSKIN DFFLRSEDLF NEMNWQKKLR AQPVLYWCAR NMSFWSSISF NLAVLMNLLV A.

Intracellular channel that mediates calcium release from the endoplasmic reticulum following stimulation by inositol 1,4,5-trisphosphate. Involved in the regulation of epithelial secretion of electrolytes and fluid through the interaction with AHCYL1. Plays a role in ER stress-induced apoptosis. Cytoplasmic calcium released from the ER triggers apoptosis by the activation of CaM kinase II, eventually leading to the activation of downstream apoptosis pathways.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IP3 Receptor 1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 4Z1B6
Concentration 0.33 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Itpr1
Gene Accession No. P11881, P29994
Gene Alias ACV; CLA4; D6Pas2; ENSMUSG00000072853; Gm10429; I145TR; inositol 1,4,5-triphosphate receptor 1; inositol 1,4,5-triphosphate receptor, type 1; inositol 1,4,5-trisphosphate receptor 1; inositol 1,4,5-trisphosphate receptor type 1; inositol 1,4,5-trisphosphate receptor, type 1; inositol 1,4,5-trisphosphate-binding protein P400; InsP>3>R type I; Insp3r; InsP3R type I; insP3R1; IP3 receptor; IP3 receptor isoform 1; Ip3r; IP-3-R; IP3R 1; IP3R1; Itpr1; Itpr-1; opisthotonus; opt; P400; Pcd6; Pcp1; Pcp-1; PPP1R94; protein PCD-6; protein phosphatase 1, regulatory subunit 94; Purkinje cell protein 1; SCA15; SCA16; SCA29; SI-SIII-SIIA; Type 1 inositol 1,4,5-trisphosphate receptor; type 1 InsP3 receptor
Gene Symbols Itpr1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 2200-2300 of human IP3R (Q14643).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16438, 25262
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.