Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IP3R 1 Rabbit anti-Mouse, Rat, Chicken, Bovine, Human, Polyclonal, MilliporeSigma™

Catalog No. ABS2129MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABS2129MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABS2129MI Supplier MilliporeSigma Supplier No. ABS2129
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect Inositol 1,4,5-trisphosphate receptor type 1 using this rabbit polyclonal Anti-IP3R1 Immunoprecipitation, and Western Blotting. ,
TRUSTED_SUSTAINABILITY

Specifications

Antigen IP3R 1
Applications Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols Itpr1,Insp3r
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to 19 amino acids from the C-terminal region of rat Inositol 1,4,5-trisphosphate receptor type 1 (IP3R1).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_001257526
Test Specificity This polyclonal antibody specifically react with IP3 Receptor 1 in multiple species, but does not recognize IP3R2 and IP3R3. It targets an epitope within 19 amino acids from the C-terminal region.
Target Species Mouse, Rat, Chicken, Bovine, Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.