Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IP3R1 Rabbit anti-Mouse, Rat, Clone: 4Z1B6, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP316414100UL
This item is not returnable.
View return policy
Description
IP3R1 Monoclonal antibody specifically detects IP3R1 in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
IP3R1 | |
Monoclonal | |
Unconjugated | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
4Z1B6 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin | |
DKFZp313E1334, DKFZp313N1434, inositol 14,5-triphosphate receptor, type 1, inositol 14,5-trisphosphate receptor type 1, INSP3R1, IP3 receptor, IP3 receptor isoform 1, IP3R, IP3R 1, IP3R1, SCA15, SCA16, spinocerebellar ataxia 15, spinocerebellar ataxia 16, Type 1 inositol 14,5-trisphosphate receptor, Type 1 InsP3 receptor | |
A synthetic peptide corresponding to a sequence within amino acids 2200-2300 of human IP3R1 (Q14643). | |
100 μg | |
Neuroscience | |
3708 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction