Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IP3R3 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP303498100UL
Description
IP3R3 Polyclonal specifically detects IP3R3 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
IP3R3 | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
3710 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunofluorescence | |
Unconjugated | |
FLJ36205, inositol 14,5-triphosphate receptor, type 3, inositol 14,5-trisphosphate receptor type 3, InsP3R3, IP3 receptor, IP3 receptor isoform 3, IP3R, IP3R 3, IP3R3, Type 3 inositol 14,5-trisphosphate receptor, Type 3 InsP3 receptor | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1630-1870 of human IP3R3 (NP_002215.2). | |
100 μL | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction