Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IP6 Kinase Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558765
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558765 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558765 Supplier Invitrogen™ Supplier No. PA558765
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CLENSEEVSQ PCQGVSVEVG KLVHKFHVGV GSLVQETLVE VGSPAEEIPE EVIQPYQEFS VEVGRLAQET SAINLLSQGI PEIDKPS Highest antigen sequence identity to the following orthologs: Mouse - 60%, Rat - 57%.

This gene encodes a dual functional inositol kinase. The encoded enzyme converts inositol hexakisphosphate to diphosphoinositol pentakisphosphate and diphosphoinositol pentakisphosphate to bis-diphosphoinositol tetrakisphosphate. This protein may be important for intracellular signaling pathways. Alternate splicing results in multiple transcript variants. A pseudogene of this gene is found on chromosome 15.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IP6 Kinase
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PPIP5K1
Gene Accession No. Q6PFW1
Gene Alias diphosphoinositol pentakisphosphate kinase 1; HISPPD2A; histidine acid phosphatase domain containing 2A; histidine acid phosphatase domain-containing protein 2A; hsVIP1; inositol hexakisphosphate and diphosphoinositol-pentakisphosphate kinase 1; Inositol pyrophosphate synthase 1; inositol-hexakisphosphate kinase; insP6 and PP-IP5 kinase 1; IP6 kinase; IP6K; IPS1; KIAA0377; PPIP5K1; VIP1; VIP1 homolog
Gene Symbols PPIP5K1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PPIP5K1. Recombinant protein control fragment (Product #RP-98141).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9677
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.