Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IP6 Kinase Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570594
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570594 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570594 Supplier Invitrogen™ Supplier No. PA570594
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IP6 Kinase Polyclonal Antibody for Western Blot

This gene encodes a dual functional inositol kinase. The encoded enzyme converts inositol hexakisphosphate to diphosphoinositol pentakisphosphate and diphosphoinositol pentakisphosphate to bis-diphosphoinositol tetrakisphosphate. This protein may be important for intracellular signaling pathways. Alternate splicing results in multiple transcript variants. A pseudogene of this gene is found on chromosome 15.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IP6 Kinase
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene PPIP5K1
Gene Accession No. Q6PFW1
Gene Alias diphosphoinositol pentakisphosphate kinase 1; HISPPD2A; histidine acid phosphatase domain containing 2A; histidine acid phosphatase domain-containing protein 2A; hsVIP1; inositol hexakisphosphate and diphosphoinositol-pentakisphosphate kinase 1; Inositol pyrophosphate synthase 1; inositol-hexakisphosphate kinase; insP6 and PP-IP5 kinase 1; IP6 kinase; IP6K; IPS1; KIAA0377; PPIP5K1; VIP1; VIP1 homolog
Gene Symbols PPIP5K1
Host Species Rabbit
Immunogen synthetic peptide directed towards the N-terminal region of human PPIP5K1.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9677
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.