Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IP6K1 Antibody, Alexa Fluor™ 750 , Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299495AF750
Description
IP6K1 Polyclonal antibody specifically detects IP6K1 in Human samples. It is validated for Western Blot, ELISA, ImmunofluorescenceSpecifications
IP6K1 | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, ELISA, Immunofluorescence | |
Alexa Fluor 750 | |
ATP:1D-myo-inositol-hexakisphosphate phosphotransferase, EC 2.7.4.21, inositol hexakisphosphate kinase 1, Inositol hexaphosphate kinase 1KIAA0263IHPK1PiUS, InsP6 kinase 1, MGC9925, Pi uptake stimulator | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human IP6K1. | |
0.1 mL | |
Neuroscience, Neurotransmission, Signal Transduction | |
9807 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction