Learn More
IP6K1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305628AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
Converts inositol hexakisphosphate (InsP6) to diphosphoinositol pentakisphosphate (InsP7/PP-InsP5). Converts 1,3,4,5,6-pentakisphosphate (InsP5) to PP-InsP4.Specifications
IP6K1 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q92551 | |
IP6K1 | |
Between 75 and 125 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunoprecipitation, Western Blot | |
Unconjugated | |
IP6K1 | |
IHPK1, PiUS, ATP:1D-myo-inositol-hexakisphosphate phosphotransferase, Pi uptake stimulator, inositol hexaphosphate kinase 1, insP6 kinase 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9807 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.