Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IP6K2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA528681
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA528681 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA528681 Supplier Invitrogen™ Supplier No. PA528681
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Predicted reactivity: Mouse (97%), Rat (97%), Xenopus laevis (82%), Rabbit (97%), Chicken (87%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The members of the inositol hexakisphosphate kinase family, IP6K1 and IP6K2, have a high affinity and selectivity for inositol hexakisphosphate (InsP6) as a substrate. IP6K1 and IP6K2 (also designated PiUS) convert InsP6 to PP-InsP5. However, neither kinase demonstrates any catalytic activity with other inositol pyrophosphates. The presence of InsP6, which inhibits serine/threonine protein phosphatases, increases the influx of calcium across the plasma membrane and implies that it may mediate the regulation of insulin exocytosis. IP6K1 was purified as a 54 kDa protein in rat brain extracts. By homology, IP6K1 and IP6K2 were characterized in mouse as a 50 kDa and 49 kDa protein, respectively. IP6K1 displays ATP synthase activity by transferring a phosphate from PP-InsP5 to ADP, which suggests a role for the IP6 kinases as high energy phosphate donors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IP6K2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.51 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene Ip6k2
Gene Accession No. Q9UHH9
Gene Alias 1500005N04Rik; ATP:1D-myo-inositol-hexakisphosphate phosphotransferase; AW050208; Ihpk2; Inositol hexakisphosphate kinase 2; inositol hexaphosphate kinase 2; inositol-hexakisphosphate kinase; insp6 kinase 2; IP6 Kinase; IP6K2; P(i)-uptake stimulator; pi uptake stimulator; Pius; TCCCIA00113
Gene Symbols Ip6k2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human IP6K2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51447
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.