Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ IP6K3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA562822
Description
Immunogen sequence: QRFYESLPLA MKRFTPQYKG TVTVHLWKDS TGHLSLVANP VKESQEPFKV STESAAVAIW QTLQQTTGSN GSDCTLAQWP HAQL Highest antigen sequence identity to the following orthologs: Mouse - 68%, Rat - 73%.
This gene encodes a protein that belongs to the inositol phosphokinase (IPK) family. This protein is likely responsible for the conversion of inositol hexakisphosphate (InsP6) to diphosphoinositol pentakisphosphate (InsP7/PP-InsP5). It may also convert 1,3,4,5,6-pentakisphosphate (InsP5) to PP-InsP4. Alternative splicing results in multiple transcript variants encoding the same protein.[provided by RefSeq, Dec 2008].
Specifications
IP6K3 | |
Polyclonal | |
Unconjugated | |
IP6K3 | |
ATP:1D-myo-inositol-hexakisphosphate phosphotransferase; D830007E07Rik; Ihpk3; Inositol hexakisphosphate kinase 3; inositol hexaphosphate kinase 3; InsP6 kinase 3; INSP6K3; IP6K3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
117283 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96PC2 | |
IP6K3 | |
Recombinant protein corresponding to Human IP6K3. Recombinant protein control fragment (Product #RP-92068). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction