Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IP6K3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5140106
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5140106 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5140106 Supplier Invitrogen™ Supplier No. PA5140106
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: LVIYDGQEPP ERAPGSPHPH EAPQAAHGSS PGGLTKVDIR MID Highest antigen sequence indentity to the following orthologs: Mouse - 42%, Rat - 49%.

This gene encodes a protein that belongs to the inositol phosphokinase (IPK) family. This protein is likely responsible for the conversion of inositol hexakisphosphate (InsP6) to diphosphoinositol pentakisphosphate (InsP7/PP-InsP5). It may also convert 1,3,4,5,6-pentakisphosphate (InsP5) to PP-InsP4. Alternative splicing results in multiple transcript variants encoding the same protein.[provided by RefSeq, Dec 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen IP6K3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene IP6K3
Gene Accession No. Q96PC2
Gene Alias ATP:1D-myo-inositol-hexakisphosphate phosphotransferase; D830007E07Rik; Ihpk3; Inositol hexakisphosphate kinase 3; inositol hexaphosphate kinase 3; InsP6 kinase 3; INSP6K3; IP6K3
Gene Symbols IP6K3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human IP6K3. Recombinant protein control fragment (Product #RP-109023).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117283
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.