Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IP6K3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5143236
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143236 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5143236 Supplier Invitrogen™ Supplier No. PA5143236
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Additional Information: This antibody is expected to recognize. Reported variants represent identical protein (NP_001136355.1, NP_473452.2. This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:128,000.

This gene encodes a protein that belongs to the inositol phosphokinase (IPK) family. This protein is likely responsible for the conversion of inositol hexakisphosphate (InsP6) to diphosphoinositol pentakisphosphate (InsP7/PP-InsP5). It may also convert 1,3,4,5,6-pentakisphosphate (InsP5) to PP-InsP4. Alternative splicing results in multiple transcript variants encoding the same protein.[provided by RefSeq, Dec 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen IP6K3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene IP6K3
Gene Accession No. Q96PC2
Gene Alias ATP:1D-myo-inositol-hexakisphosphate phosphotransferase; D830007E07Rik; Ihpk3; Inositol hexakisphosphate kinase 3; inositol hexaphosphate kinase 3; InsP6 kinase 3; INSP6K3; IP6K3
Gene Symbols IP6K3
Host Species Goat
Immunogen Peptide with sequence C-KESQEPFKVSTES.
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117283
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.