Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ IP7e
GREENER_CHOICE

Catalog No. 569910
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
569910 10 mg
569950 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 569910 Supplier Tocris Bioscience™ Supplier No. 5699/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent Nurr1 activator

IP7e is a potent Nurr1 activator (EC50 = 3.9 nM). Delays onset and reduces severity of symptoms in mice with experimental autoimmune encephalomyelitis (EAE). Also suppresses NF-κB signaling. Brain penetrant and orally bioavailable.

Specifications

Chemical Name or Material 6-[4-[(2-Methoxyethoxy)methyl]phenyl]-5-methyl-3-phenyl-isoxazolo[4,5-c]pyridin-4(5H)-one
CAS 500164-74-9
Quantity 10 mg
Target NFkB/IkB Inhibitors
Molecular Formula C23H22N2O4
Formula Weight 390.43
Purity >98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.