Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ IPA 3
GREENER_CHOICESDP

Catalog No. 362250R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36-225-0R 50 mg
36-221-0R 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 36-225-0R Supplier Tocris Bioscience™ Supplier No. 3622/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Group I PAK inhibitor

IPA 3 is a group I p21-activated kinase (PAK) inhibitor (IC50 = 2.5 μM at PAK1). Targets the autoregulatory mechanism and promotes the inactive conformation of PAKs. Inhibits PAK1-mediated signaling in vivo; potential anti-tumor agent.

Negative Control also available.

Specifications

Quantity 50 mg
Product Type IPA 3
Purity >96%
Molecular Weight (g/mol) 350.45
Inhibitors IPA 3

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.