Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IPAD Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144526
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144526 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144526 Supplier Invitrogen™ Supplier No. PA5144526
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IPAD Polyclonal Antibody for Western Blot, ELISA

Required for bacterial invasion of host cells. Controls IpaB and IpaC secretion, and the efficiency with which they are physically inserted into target cell membranes. These proteins are exported via T3SS to form a pore in the host membrane that allows the translocation of the other effectors into the host cytoplasm. Along with IpaB, is essential for both blocking secretion through the Mxi/Spa translocon in the absence of a secretion-inducing signal, and for controlling the level of secretion in the presence of this signal.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IPAD
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3.819 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene ipaD
Gene Accession No. P18013
Gene Alias Invasin IpaDipaD CP0126
Gene Symbols ipaD
Host Species Rabbit
Immunogen Recombinant Shigella flexneri IPAD protein (1-332aa).
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1238053
Target Species Bacteria
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.