Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ IPAD Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144413
Description
IPAD Polyclonal Antibody for Western Blot, ELISA
Required for bacterial invasion of host cells. Controls IpaB and IpaC secretion, and the efficiency with which they are physically inserted into target cell membranes. These proteins are exported via T3SS to form a pore in the host membrane that allows the translocation of the other effectors into the host cytoplasm. Along with IpaB, is essential for both blocking secretion through the Mxi/Spa translocon in the absence of a secretion-inducing signal, and for controlling the level of secretion in the presence of this signal.
Specifications
IPAD | |
Polyclonal | |
Unconjugated | |
ipaD | |
Invasin IpaDipaD CP0126 | |
Rabbit | |
Protein G | |
RUO | |
1238053 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Western Blot | |
3.819 mg/mL | |
PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
P18013 | |
ipaD | |
Recombinant Shigella flexneri IPAD protein (1-332aa). | |
100 μL | |
Primary | |
Bacteria | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction