Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IPAG, Tocris Bioscience™

Catalog No. 054450
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent σ antagonist

Potent σ-receptor antagonist.

Chemical Identifiers

CAS 193527-91-2
Molecular Formula C17H22IN3
Molecular Weight (g/mol) 395.288
InChI Key UUKPIWYXWLJPJF-UHFFFAOYSA-N
Synonym ipag, 1-4-iodophenyl-3-2-adamantyl guanidine, 2-2-adamantyl-1-4-iodophenyl guanidine, tocris-0544
PubChem CID 4239764
IUPAC Name 2-(2-adamantyl)-1-(4-iodophenyl)guanidine
SMILES C1C2CC3CC1CC(C2)C3N=C(N)NC4=CC=C(C=C4)I

Specifications

Quantity 50 mg
Formula Weight 395.29
Percent Purity >98%
Chemical Name or Material IPAG
Recommended Storage Store at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.