Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IPCEF1 Antibody (OTI3G2), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271781G
Description
IPCEF1 Monoclonal specifically detects IPCEF1 in Human samples. It is validated for Western Blot.Specifications
IPCEF1 | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
IPCEF1 | |
Full length recombinant protein of human IPCEF1 (NP_056368) produced in HEK293T cells. | |
0.1 mL | |
26034.0 | |
Store at 4°C in the dark. |
Western Blot | |
OTI3G2 | |
Western Blot | |
interaction protein for cytohesin exchange factors 1, interactor protein for cytohesin exchange factors 1, KIAA0403interaction protein for cytohesin exchange factors 1 Interaction protein forcytohesin exchange factors 1, phosphoinositide binding protein PIP3-E, Phosphoinositide-binding protein PIP3-E, PIP3-E, RP3-402L9.2 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction