Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IPCEF1 Monoclonal Antibody (OTI3G2), TrueMAB™, OriGene
SDP

Catalog No. 502539818
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-253-9818 30 μL
50-167-5771 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-253-9818 Supplier OriGene Supplier No. TA505252S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IPCEF1 Monoclonal antibody specifically detects IPCEF1 in Human samples. It is validated for Western Blot

Specifications

Antigen IPCEF1
Applications Western Blot
Classification Monoclonal
Clone OTI3G2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene FATE1
Gene Accession No. Q969F0
Gene Alias A130090K04Rik; interaction protein for cytohesin exchange factors 1; interaction protein for cytohesin exchange factors 1 Interaction protein for cytohesin exchange factors 1; interactor protein for cytohesin exchange factors 1; Ipcef1; KIAA0403; phosphoinositide binding protein PIP3-E; phosphoinositide-binding protein PIP3-E; PIP3-E; RP3-402L9.2
Host Species Mouse
Immunogen Full length human recombinant protein of human FATE1 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 89885
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.