Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IPCEF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583028
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583028 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583028 Supplier Invitrogen™ Supplier No. PA583028
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VERASECKKK HAFKISHPQI KTFYFAAENV QEMNVWLNKL GSAVIHQEST TKDEECYSES EQEDPEIAAE T Highest antigen sequence indentity to the following orthologs: Rat - 86%, Mouse - 86%.

Enhances the promotion of guanine-nucleotide exchange by PSCD2 on ARF6 in a concentration-dependent manner. Target protein binds guanine nucleotide exchange factor cytohesin 2 and increases cytohesin 2 mediated formation of ADP-ribosylation factor GTP-ase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IPCEF1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene IPCEF1
Gene Accession No. Q8WWN9
Gene Alias A130090K04Rik; interaction protein for cytohesin exchange factors 1; interaction protein for cytohesin exchange factors 1 Interaction protein for cytohesin exchange factors 1; interactor protein for cytohesin exchange factors 1; Ipcef1; KIAA0403; phosphoinositide binding protein PIP3-E; phosphoinositide-binding protein PIP3-E; PIP3-E; RP3-402L9.2
Gene Symbols IPCEF1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human IPCEF1. Recombinant protein control fragment (Product #RP-108322).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26034
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.