Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva IPG Buffers

Catalog No. p-3667666 Shop All Cytiva Products
Change view
Click to view available options
pH Range:
3 to 10
3 to 10 nonlinear
3 to 11 nonlinear
4 to 7
6 to 11
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. pH Range
45-000-352 6 to 11
45-000-343 4 to 7
45-000-344 3 to 10
45-000-345 3 to 10 nonlinear
45-000-408 3 to 11 nonlinear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 45-000-352 Supplier Cytiva Supplier No. 17600178
Only null left
Add to Cart
Add to Cart

Produce more uniform conductivity along the Immobiline™ DryStrip during focusing. Cytiva IPG Buffers are ampholyte-containing buffers formulated for use with Immobiline™ DryStrip gels.

  • Eliminates background staining
  • Use with Immobiline DryStrip gels to improve protein solubility
  • Ensures optimal conditions for IEF separation
  • High sample loading capacity, improved resolution, and increases the probability of detecting low abundance proteins when used with Immobiline DryStrip gels

Order Info

Supplied in 1mL aliquots of water; order Immobiline Drystrip gels separately.

TRUSTED_SUSTAINABILITY

Specifications

Color Colorless to Light Yellow
pH Range 6 to 11
Form Liquid
For Use With (Application) IEF, 2-D Electrophoresis
Quantity 1mL
Storage Requirements 2° to 8°C
Product Type IPG Buffer
For Use With (Equipment) Immobiline DryStrip Gels
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.