Learn More
Cytiva IPGbox
Catalog No. 09928081 Shop All Cytiva Products
Change view
Click to view available options
No. of Gels:
Up to 12 Immobiline Dry Strip Gel
Catalog No. No. of Gels
09-928-081 Up to 12 Immobiline Dry Strip Gel
1 options
Catalog No. 09-928-081 Supplier Cytiva Supplier No. 28933465
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart
Includes
IPGbox: 1 IPGbox, 1 IPGbox Kit (10 Reswell trays, 1 IPGbox insert, instructions)
Tools for enhancing reswelling of precast Immobiline DryStrip gels (IPG strips)
  • Oil free rehydratation of up to 12 Immobiline DryStrip gels or other IPG strips independently and simultaneously
  • Lid protects IPG strips from dust and contamination
  • Load samples for IEF during rehydration by including them in the rehydration bufferor load sample using sample cups
  • IPGbox used together with IPGbox Kit (Cat. No 45002627) to rehydrate 7 to 24cm Immobiline DryStrip gels
  • Includes reswelling tray made of hydrophobic material to prevent sticking of rehydration solution
  • IPGbox Insert located within lid of IPGbox to maintain optimal conditions for rehydration
TRUSTED_SUSTAINABILITY
Specifications
Type IPGBox
Includes 1 IPGbox and 1 IPGbox kit
No. of Gels Up to 12 Immobiline Dry Strip Gel