Learn More
Cytiva IPGbox Immobiline™ DryStrip Gel Box
Catalog No. 45002630 Shop All Cytiva Products
Change view
Click to view available options
Type:
Gel Box
Catalog No. Type
45-002-630 Gel Box
1 options
Catalog No. 45-002-630 Supplier Cytiva Supplier No. 28938445
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Specifications
Description IPGbox Immobiline DryStrip Gel Box
For Use With (Application) Enhances the reswelling of precast Immobiline DryStrip gels (IPG strips)
Type Gel Box