Learn More
Cytiva IPGbox Kit
Catalog No. 45002627 Shop All Cytiva Products
Change view
Click to view available options
Material:
PET
Catalog No. Material
45-002-627 PET
1 options
Catalog No. 45-002-627 Supplier Cytiva Supplier No. 28933492
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart
Includes
IPGbox Kit: 10 Reswell trays, 1 IPGbox insert, instructions
Tools for enhancing reswelling of precast Immobiline DryStrip gels (IPG strips)
  • Oil free rehydratation of up to 12 Immobiline DryStrip gels or other IPG strips independently and simultaneously
  • Lid protects IPG strips from dust and contamination
  • Load samples for IEF during rehydration by including them in rehydration bufferor load sample using sample cups
  • IPGbox (Cat. No. 45002631) used together with IPGbox Kit to rehydrate 7 to 24cm Immobiline DryStrip gels
  • Includes reswelling tray made of hydrophobic material to prevent sticking of rehydration solution
  • IPGbox Insert located within lid of IPGbox to maintain optimal conditions for rehydration
TRUSTED_SUSTAINABILITY
Specifications
Type IPGBox Kit
Includes 10 Reswell trays and 1 IPGbox insert
Material PET