Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Ipilimumab Recombinant Human Monoclonal Antibody

Human Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA541799
This item is not returnable.
View return policy
Description
Endotoxin level < 0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.
Ipilimumab (Yervoy) is a humanized monoclonal antibody that is approved by the U.S. Food and Drug Administration for the treatment of patients with unresectable or metastatic melanoma. Ipilimumab works to activate the immune system by targeting CTLA-4 (Cytotoxic T-lymphocyte protein 4), a negative regulator of cytotoxic T-lymphocyte activity. Ipilimumab binds to CTLA-4, blocking the inhibitory signal, which allows the cytotoxic T-lymphocytes to destroy the cancer cells.
Specifications
| Ipilimumab | |
| Recombinant Monoclonal | |
| Unconjugated | |
| BMS-734016; MDX-010; MDX-CTLA-4; Yervoy | |
| Human CTLA4/CD152. | |
| 100 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid | |
| Chemical |
| ELISA, SDS-Page | |
| 2.87 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Human | |
| Protein A/G | |
| RUO | |
| Human | |
| Antibody | |
| IgG1 κ |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction