Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IPMK Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111456
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111456 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111456 Supplier Invitrogen™ Supplier No. PA5111456
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EYNNNFHVLS STANGKIESS VGKSLSKMYA RHRKIYTKKH HSQTSLKVEN LEQDNGWKSM SQEHLNGNVL SQLEKVFYHL PTGC Highest antigen sequence indentity to the following orthologs: Mouse - 75%, Rat - 81%.

This gene encodes a member of the inositol phosphokinase family. The encoded protein has 3-kinase, 5-kinase and 6-kinase activities on phosphorylated inositol substrates. The encoded protein plays an important role in the biosynthesis of inositol 1, 3, 4, 5, 6-pentakisphosphate, and has a preferred 5-kinase activity. This gene may play a role in nuclear mRNA export. Pseudogenes of this gene are located on the long arm of chromosome 13 and the short arm of chromosome 19.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IPMK
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene IPMK
Gene Accession No. Q8NFU5
Gene Alias 2410017C19Rik; AA408208; Impk; Inositol 1,3,4,6-tetrakisphosphate 5-kinase; inositol polyphosphate kinase 2; inositol polyphosphate multikinase; Ipk2; Ipmk; LOW QUALITY PROTEIN: inositol polyphosphate multikinase
Gene Symbols IPMK
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human IPMK. Recombinant protein control fragment (Product #RP-106783).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 253430
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.