Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IPO-38 Antibody (IPO-38), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP2451940.1MG
Description
Ensure accurate, reproducible results in Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence
IPO-38 Monoclonal specifically detects IPO-38 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
IPO-38 | |
Monoclonal | |
0.2mg/mL | |
Flow Cytometry 5 - 10 ul/million cells in 0.1ml, Immunohistochemistry-Paraffin 1:25 - 1:50, Immunofluorescence 1:25 - 1:50 | |
Mouse | |
14.5 kDa | |
0.1 mg | |
Primary | |
Human, Mouse, Rat | |
Purified |
Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence | |
IPO-38 | |
Unconjugated | |
10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
Spleen cells of a patient with hairy cell leukemia | |
Protein A or G purified | |
RUO | |
Recognizes a protein of 14-16kDa, which is a novel nuclear antigen of proliferating cells. IPO-38 antigen is present in the nuclei of proliferating cells such as Hodgkin s disease and non-Hodgkin s lymphomas, different forms of leukemias, breast and colorectal carcinomas, and PHA-stimulated lymphocytes. It is not expressed in the cells of non-stimulated lymphocytes and granulocytes. IPO-38 may be a useful marker of cell proliferation during monitoring of tumor progression. | |
Store at 4C. | |
IgM κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction