Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IPP Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | IPP |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
IPP Polyclonal specifically detects IPP in Human samples. It is validated for Western Blot.Specifications
| IPP | |
| Polyclonal | |
| Rabbit | |
| Q9Y573 | |
| 3652 | |
| Synthetic peptides corresponding to IPP(intracisternal A particle-promoted polypeptide) The peptide sequence was selected from the C terminal of IPP (NP_005888). Peptide sequence EMQGLIYVIGGISNEGIELRSFEVYDPLSKRWSPLPPMGTRRAYLGVAAL. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| actin-binding protein IPP, intracisternal A particle-promoted polypeptideKLHL27Kelch-like protein 27 | |
| IPP | |
| IgG | |
| 65 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title