Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IPP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5140178
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5140178 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5140178 Supplier Invitrogen™ Supplier No. PA5140178
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: SGLGVTVLGG MVYAIGGEKD SMIFDCTECY DPVTKQWTTV ASMNHPRCGL GVCVCYGAIY ALGGWVGAEI GNTIERFDPD Highest antigen sequence indentity to the following orthologs: Mouse - 96%, Rat - 96%.

The protein encoded by this gene is a member of the kelch family of proteins, which is characterized by a 50 amino acid repeat which interacts with actin. Transcript variants have been described but their full-length nature has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IPP
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene IPP
Gene Accession No. Q9Y573
Gene Alias Actin-binding protein IPP; IAP promoted placental; intracisternal A particle-promoted polypeptide; IPP; kelch-like family member 27; kelch-like protein 27; KLHL27
Gene Symbols IPP
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human IPP. Recombinant protein control fragment (Product #RP-109486).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3652
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.