Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IPPK Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554243
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554243 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554243 Supplier Invitrogen™ Supplier No. PA554243
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GCLLYKTLQV QMLDLLDIEG LYPLYNRVER YLEEFPEERK TLQIDGPYDE AFYQKLLDLS TEDDGTVAFA LTKVQQYR Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 96%.

The protein encoded by this gene is a kinase that phosphorylates position 2 of inositol-1, 3, 4, 5, 6-pentakisphosphate to form inositol-1, 2, 3, 4, 5, 6-hexakisphosphate (InsP6). InsP6 has a variety of functions, including stimulation of DNA repair, endocytosis, and mRNA export.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IPPK
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene IPPK
Gene Accession No. Q5PXE9, Q6P1C1, Q9H8X2
Gene Alias 1,3,4,5,6-pentakisphosphate 2-kinase; 1810043M15Rik; bA476B13.1; bA476B13.1 (novel protein); C9orf12; FLJ13163; hypothetical protein LOC541511; inositol 1,3,4,5,6-pentakisphosphate 2-kinase; inositol 1,3,4,5,6-pentakisphosphate 2-kinase, like; inositol polyphosphate kinase 1; inositol-1,3,4,5,6-pentakisphosphate 2-kinase; Inositol-pentakisphosphate 2-kinase; ins(1,3,4,5,6)P5 2-kinase; InsP5 2-kinase; INSP5K2; InsP6; IP5K; IPK1; IPK1 homolog; ipk1l; ippk; KIAA0699; RGD1311271; rIpk1; si:dkey-42i9.3; zgc:110769
Gene Symbols IPPK
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human IPPK. Recombinant protein control fragment (Product #RP-92947).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 306808, 64768, 75678
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.