Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ IPPK Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA599642
Description
Antibody detects endogenous levels of total InsP5 2-kinase.
The protein encoded by this gene is a kinase that phosphorylates position 2 of inositol-1, 3, 4, 5, 6-pentakisphosphate to form inositol-1, 2, 3, 4, 5, 6-hexakisphosphate (InsP6). InsP6 has a variety of functions, including stimulation of DNA repair, endocytosis, and mRNA export.
Specifications
IPPK | |
Polyclonal | |
Unconjugated | |
IPPK | |
1,3,4,5,6-pentakisphosphate 2-kinase; 1810043M15Rik; bA476B13.1; bA476B13.1 (novel protein); C9orf12; FLJ13163; hypothetical protein LOC541511; inositol 1,3,4,5,6-pentakisphosphate 2-kinase; inositol 1,3,4,5,6-pentakisphosphate 2-kinase, like; inositol polyphosphate kinase 1; inositol-1,3,4,5,6-pentakisphosphate 2-kinase; Inositol-pentakisphosphate 2-kinase; ins(1,3,4,5,6)P5 2-kinase; InsP5 2-kinase; INSP5K2; InsP6; IP5K; IPK1; IPK1 homolog; ipk1l; ippk; KIAA0699; RGD1311271; rIpk1; si:dkey-42i9.3; zgc:110769 | |
Rabbit | |
Affinity chromatography | |
RUO | |
64768, 75678 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q6P1C1, Q9H8X2 | |
IPPK | |
A synthesized peptide derived from human IPPK(Accession Q9H8X2), corresponding to amino acid residues G15-E65. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction