Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ipratropium Bromide Monohydrate 98.0+%, TCI America™
SDP

Catalog No. I09071G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I09071G 1 g
I0907200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I09071G Supplier TCI America Supplier No. I09071G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 66985-17-9
Molecular Formula C20H32BrNO4
Molecular Weight (g/mol) 430.383
MDL Number MFCD09842549
InChI Key KEWHKYJURDBRMN-IUZMIPGZSA-M
Synonym Atropine Isopropyl Bromide
PubChem CID 129318280
IUPAC Name [(1R,5S)-8-methyl-8-propan-2-yl-8-azoniabicyclo[3.2.1]octan-3-yl] (2R)-3-hydroxy-2-phenylpropanoate;bromide;hydrate
SMILES CC(C)[N+]1(C2CCC1CC(C2)OC(=O)C(CO)C3=CC=CC=C3)C.O.[Br-]

Specifications

Melting Point 231°C
Color White
Quantity 1 g
Formula Weight 412.37
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Ipratropium Bromide Monohydrate
RTECSNumber YM3700000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.