Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ipratropium Bromide, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18603506 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-603-506 5 g Amber Glass Bottle
18-603-505 1 g Amber Glass Bottle
18-603-507 25 g Amber Glass Bottle
18-603-508 100 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-603-506 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. I11785GM
Add to Cart
Add to Cart

I1178, 66985-17-9, C20H30BrNO3·H2O

Spectrum™ Chemical Ipratropium Bromide, USP is a bronchodilator that dilates airways in the lungs. All Spectrum Chemical manufactured USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 66985-17-9
Molecular Formula C20H32BrNO4
Molecular Weight (g/mol) 430.38
InChI Key KEWHKYJURDBRMN-XFQAGIBXSA-M
IUPAC Name (1R,5S)-3-[(3-hydroxy-2-phenylpropanoyl)oxy]-8-methyl-8-(propan-2-yl)-8-azabicyclo[3.2.1]octan-8-ium hydrate bromide
SMILES O.[Br-].CC(C)[N+]1(C)[C@H]2CC[C@@H]1CC(C2)OC(=O)C(CO)C1=CC=CC=C1

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Packaging Amber Glass Bottle
Quantity 5 g
Percent Purity 98 to 102%
Grade USP
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Ipratropium Bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.