Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ipriflavone 98.0+%, TCI America™
SDP

Catalog No. I066925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I066925G 25 g
I06695G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I066925G Supplier TCI America Supplier No. I066925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 35212-22-7
Molecular Formula C18H16O3
Molecular Weight (g/mol) 280.32
MDL Number MFCD00221719
InChI Key SFBODOKJTYAUCM-UHFFFAOYSA-N
Synonym ipriflavone, osten, yambolap, osteofix, 7-isopropoxyisoflavone, iprosten, ipriflavonum, ipriflavona, osteoquine, quinogin
PubChem CID 3747
ChEBI CHEBI:31719
IUPAC Name 3-phenyl-7-(propan-2-yloxy)-4H-chromen-4-one
SMILES CC(C)OC1=CC=C2C(=O)C(=COC2=C1)C1=CC=CC=C1

Specifications

Melting Point 117°C
Color White
Quantity 25 g
Formula Weight 280.32
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ipriflavone
RTECSNumber DJ3100500
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.