Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ IPTG
GREENER_CHOICESDP

Catalog No. 550910G Shop All Novus Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
550910G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 550910G Supplier R&D Systems™ Supplier No. 5509/10G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Used in cloning procedures with X-GAL

IPTG is an used in combination with X-GAL (Cat. No. 5510) in cloning procedures requiring expression of β-galactosidase activity. Non-metabolizable analog of galactose.

This product has <0.1% dioxane content.

Specifications

Chemical Name or Material 1-Methylethyl 1-thio-β-D-galactopyranoside
CAS 367-93-1
Synonym 1-Methylethyl 1-thio-β-D-galactopyranoside
Quantity 10 g
Solubility Soluble to 100mM in water
Product Line Tocris
Target Buffers, Chelators and Reagents
Molecular Formula C9H18O5S
Formula Weight 238.3
Physical Form Solid
Product Type Bioactive Small Molecule
Purity 99%
Show More Show Less

Research Use Only. Not For Human or Veterinary Use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.