Learn More
Description
- Four-button placket
- Rib knit henley collar
- Raglan sleeves
- Concealed rib knit cuffs
- Full underarm gusset
- Concealed chest pocket with pencil stall
- Concealed pencil stall sleeve pocket
- Contrasting thread color iQ Series™ seam tag Exterior CAT 2 2112 labels
- Fabric: Exclusively with Westex G2™ fabrics by Milliken™ - Flame-resistant, 5.3 oz. (180 g/m2) 69% Cotton / 25% Polyester / 6% Polyoxadiazole
- Care: Home Wash
- Packaging: iQ Series™ hang tag /Polybag
- Protection: Arc Rating ATPV 8.2 calories/cm2
- Country of Origin: Imported
Specifications
Specifications
| Size | 3X-Large |
| Gender | Men's |
| Material | Cotton, Polyester, POD |
| Color | Charcoal |
| Sleeve Style | Long Sleeve |
| Arc Flash Rating | ATPV 8.2 calories/cm2 |
| Certifications/Compliance | NFPA 2112, Standard on Flame- Resistant Garments for Protection of Industrial Personnel Against Flash Fire, 2018 Edition, EN ISO 11612:2015, Protective clothing against heat and flame; EN 340:2003, Protective clothing - General requirements; IEC 61482- 2:2009, Live working - Protective clothing against the thermal hazards of an electric arc |
| Size (English) Chest | 54 to 58.5 in. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.