Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bulwark™ FR IQ Series™ Comfort Knit Long Sleeve Henley Tall

Catalog No. 171501161 Shop All Red Kap Industries Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large
Size (English) Chest:
35 to 38 in.
38 to 41 in.
41 to 45 in.
45 to 49.5 in.
49.5 to 54 in.
54 to 58.5 in.
58.5 to 63 in.
63 to 67.5 in.
67.5 to 72 in.
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Size (English) Chest
17-150-1161 Large 41 to 45 in.
17-150-1157 Small 35 to 38 in.
17-150-1159 Medium 38 to 41 in.
17-150-1163 X-Large 45 to 49.5 in.
17-150-1165 2X-Large 49.5 to 54 in.
17-150-1167 3X-Large 54 to 58.5 in.
17-150-1169 4X-Large 58.5 to 63 in.
17-150-1171 5X-Large 63 to 67.5 in.
17-150-1173 6X-Large 67.5 to 72 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 17-150-1161 Supplier Bulwark™ FR Supplier No. QT20CHLGLN

Bulwark™ flame resistant long sleeve knit shirt.

  • Four-button placket
  • Rib knit henley collar
  • Raglan sleeves
  • Concealed rib knit cuffs
  • Full underarm gusset
  • Concealed chest pocket with pencil stall
  • Concealed pencil stall sleeve pocket
  • Contrasting thread color iQ Series™ seam tag Exterior CAT 2 2112 labels
  • Fabric: Exclusively with Westex G2™ fabrics by Milliken™ - Flame-resistant, 5.3 oz. (180 g/m2) 69% Cotton / 25% Polyester / 6% Polyoxadiazole
  • Care: Home Wash
  • Packaging: iQ Series™ hang tag /Polybag
  • Protection: Arc Rating ATPV 8.2 calories/cm2
  • Country of Origin: Imported

Specifications

Size Large
Gender Men's
Material Cotton, Polyester, POD
Color Charcoal
Sleeve Style Long Sleeve
Arc Flash Rating ATPV 8.2 calories/cm2
Certifications/Compliance NFPA 2112, Standard on Flame- Resistant Garments for Protection of Industrial Personnel Against Flash Fire, 2018 Edition, EN ISO 11612:2015, Protective clothing against heat and flame; EN 340:2003, Protective clothing - General requirements; IEC 61482- 2:2009, Live working - Protective clothing against the thermal hazards of an electric arc
Size (English) Chest 41 to 45 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.