Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bulwark™ FR IQ Series™ Comfort Knit Long Sleeve Henley Tall
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large
Size (English) Chest:
35 to 38 in.
38 to 41 in.
41 to 45 in.
45 to 49.5 in.
49.5 to 54 in.
54 to 58.5 in.
58.5 to 63 in.
63 to 67.5 in.
67.5 to 72 in.
Description
- Four-button placket
- Rib knit henley collar
- Raglan sleeves
- Concealed rib knit cuffs
- Full underarm gusset
- Concealed chest pocket with pencil stall
- Concealed pencil stall sleeve pocket
- Contrasting thread color iQ Series™ seam tag Exterior CAT 2 2112 labels
- Fabric: Exclusively with Westex G2™ fabrics by Milliken™ - Flame-resistant, 5.3 oz. (180 g/m2) 69% Cotton / 25% Polyester / 6% Polyoxadiazole
- Care: Home Wash
- Packaging: iQ Series™ hang tag /Polybag
- Protection: Arc Rating ATPV 8.2 calories/cm2
- Country of Origin: Imported
Specifications
Specifications
Size | 3X-Large |
Gender | Men's |
Material | Cotton, Polyester, POD |
Color | Charcoal |
Sleeve Style | Long Sleeve |
Arc Flash Rating | ATPV 8.2 calories/cm2 |
Certifications/Compliance | NFPA 2112, Standard on Flame- Resistant Garments for Protection of Industrial Personnel Against Flash Fire, 2018 Edition, EN ISO 11612:2015, Protective clothing against heat and flame; EN 340:2003, Protective clothing - General requirements; IEC 61482- 2:2009, Live working - Protective clothing against the thermal hazards of an electric arc |
Size (English) Chest | 54 to 58.5 in. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction