Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bulwark™ FR IQ Series™ Long Sleeve Comfort Woven Lightweight Shirt Tall

Catalog No. 171501155 Shop All Red Kap Industries Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large
Length (English) Sleeve:
36.25 in.
36.75 in.
Size (English) Chest:
34.5 to 38 in.
38.5 to 42 in.
42.5 to 46 in.
46.5 to 50 in.
50.5 to 54 in.
54.5 to 58 in.
58.5 to 62 in.
62.5 to 66 in.
66.5 to 70 in.
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Length (English) Sleeve Size (English) Chest
17-150-1155 6X-Large 36.75 in. 66.5 to 70 in.
17-150-1139 Small 36.25 in. 34.5 to 38 in.
17-150-1141 Medium 36.25 in. 38.5 to 42 in.
17-150-1143 Large 36.75 in. 42.5 to 46 in.
17-150-1145 X-Large 36.75 in. 46.5 to 50 in.
17-150-1147 2X-Large 36.75 in. 50.5 to 54 in.
17-150-1149 3X-Large 36.75 in. 54.5 to 58 in.
17-150-1151 4X-Large 36.75 in. 58.5 to 62 in.
17-150-1153 5X-Large 36.75 in. 62.5 to 66 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 17-150-1155 Supplier Bulwark™ FR Supplier No. QS50NV6XLN
Add to Cart
Add to Cart

Flame resistant long sleeve comfort lightweight regular shirt

  • Fabric: Bulwark Exclusive 5.5 oz. 50% Aramid 25% Lyocell 25% Modacrylic- Flame-resistant
  • Care: Industrial Laundry - Light Soil, Home Wash
  • Banded, Topstitched collar with red trim at neck seam
  • Topstitched cuffs with two-button adjustable closures
  • Tailored sleeve placket
  • Hemmed front placket with button closure
  • Larger patch chest pockets
  • Full side seam gusset
  • iQ Series™ seam tag
  • Packaging:
    • iQ Series™ hang tag
    • Polybag
    • Hanger with size clip
    • Protection: Arc Rating ATPV 8.5 calories/cm2;
    • Country of Origin: Imported

    Specifications

    Size 6X-Large
    Gender Men's
    Material Aramid, Lyocell, Modacrylic
    Color Navy
    Neck Style Collar
    Closure Type Button
    No. of Pockets 2
    Arc Flash Rating ATPV 8.5 calories/cm2
    Certifications/Compliance NFPA 2112, Standard on Flame- Resistant Garments for Protection of Industrial Personnel Against Flash Fire, 2018 Edition, EN ISO 11612:2015, Protective clothing against heat and flame; EN 340:2003, Protective clothing - General requirements; IEC 61482- 2:2009, Live working - Protective clothing against the thermal hazards of an electric arc
    Size (English) Chest 66.5 to 70 in.
    Length (English) Sleeve 36.75 in.
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.