Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bulwark™ iQ Series™ Men's Comfort Knit Black Flame Resistant Henley
Click to view available options
Size:
2X-Large
2X-Large (Long)
3X-Large
3X-Large (Long)
4X-Large
Large
Large (Long)
Medium
Small
X-Large
X-Large (Long)
Description
- One concealed chest pocket with pencil stall and one concealed pencil stall sleeve pocket
- Hidden rib knit cuffs to keep sleeves in place
- Moisture-wicking, breathable fabric
- Four-button placket
- Hazard: Flash Fire, Arc Flash
- Wash Care: Home Wash
- Fabric: Exclusively with Westex G2™ fabrics by Milliken™ - Flame resistant, 5.3 oz. (180 g/m2)
- Blend: 69% Cotton/ 25% Polyester/ 6% Polyoxadiazole
- Packaging: Polybag
Specifications
Specifications
Size | Small |
Gender | Men's |
Material | Cotton, Polyester, POD |
Color | Black |
Neck Style | Henley |
Sleeve Style | Raglan Sleeve |
Construction | Knit |
Closure Type | Button |
Disposable | Reusable |
No. of Pockets | 2 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction