Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bulwark™ iQ Series™ Men's Comfort Knit Red Flame Resistant Henley
Click to view available options
Size:
2X-Large
2X-Large (Long)
3X-Large
3X-Large (Long)
4X-Large
Large
Large (Long)
Medium
Small
X-Large
X-Large (Long)
Description
- With raglan sleeves and underarm gussets for added mobility, so you can bend, reach, stretch and work better
- One concealed chest pocket with pencil stall and one concealed pencil stall sleeve pocket
- Hidden rib knit cuffs to keep sleeves in place
- Moisture-wicking, breathable fabric
- Certifications Level 1: ASTM F1506, NFPA 2112, NFPA 70E
- Certifications Level 2: Category 2 Protection, NFPA 2112 - 2018 Compliant
- Hazard: Flash Fire, Arc Flash
- Wash Care: Home Wash
- Fabric: Exclusively with Westex G2™ fabrics by Milliken™ - Flame resistant, 5.3 oz. (180 g/m2)
- Blend: 69% Cotton/25% Polyester/6% Polyoxadiazole
- Closure: Four-button placket
- Product Collection: iQ Series Comfort, iQ Series
- Packaging: Polybag
Specifications
Specifications
Size | 3X-Large |
Gender | Men's |
Material | Cotton, Polyester, POD |
Color | Red |
Neck Style | Henley |
Sleeve Style | Raglan Sleeve |
Construction | Knit |
Closure Type | Button |
Disposable | Reusable |
No. of Pockets | 2 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction