Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bulwark™ iQ Series™ Men's Lightweight Navy Flame Resistant Ripstop Pant

Catalog No. 17204435
Click to view available options
Size:
28 x 37 in.
30 x 30 in.
30 x 32 in.
30 x 37 in.
32 x 30 in.
32 x 32 in.
32 x 34 in.
34 x 30 in.
34 x 32 in.
34 x 34 in.
34 x 37 in.
36 x 30 in.
36 x 32 in.
36 x 34 in.
36 x 37 in.
38 x 30 in.
38 x 32 in.
38 x 34 in.
38 x 37 in.
40 x 30 in.
40 x 32 in.
40 x 34 in.
40 x 37 in.
42 x 30 in.
42 x 32 in.
42 x 34 in.
42 x 37 in.
44 x 30 in.
44 x 37 in.
46 x 37 in.
48 x 37 in.
50 x 37 in.
52 x 37 in.
54 x 37 in.
56 x 37 in.

iQ Series™ Men's Lightweight Navy Flame Resistant Ripstop Pant is stylishly designed to keep you cool and in command all day long. It is NFPA™ 2112 compliant, with a straight-fit, modern jean style.

  • Sits at the waist, regular in the seat and thigh with a slightly tapered leg
  • Jean style cut with one-piece waistband with concealed button closure
  • Wide/long belt loops to fit larger belts
  • Full crotch gusset and double articulated knees for ease of movement and increased mobility
  • Two scoop front pockets and one concealed pocket on right pocket bag, two patch hip pockets, one cargo pocket with snap closures on left leg and one double utility pocket on right leg
  • Hazard: Flash Fire, Arc Flash
  • Wash Care: Industrial Laundry - Heavy Soil, Industrial Laundry - Light Soil, Home Wash
  • Fabric: Flame resistant, 6.9 oz (235 g/m2) Ripstop Twill
  • Blend: 65% Cotton/35% Polyester
  • Country of Origin: Imported, Made in USA Fabric

Specifications

Size 32 x 32 in.
Gender Men's
Material Cotton, Polyester
Color Navy
Seam Type Stitched
No. of Pockets 7
Closure Type Concealed Button
Disposable Reusable
Hazard Risk Category Category 2
Arc Flash Rating 9
Certifications/Compliance ASTM F1506, NFPA 2112, NFPA 70E
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.